Product Details

SNP ID
rs1064574
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1577610 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAAGGAGAGCGGCCTCCTGGGCA[C/T]GCCACTCAGGCACGGTGAGGCTCTG
Phenotype
MIM: 603573 MIM: 611009
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MBD3 PubMed Links

Gene Details

Gene
MBD3
Gene Name
methyl-CpG binding domain protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281453.1 1560 UTR 3 NP_001268382.1
NM_001281454.1 1560 UTR 3 NP_001268383.1
Gene
MEX3D
Gene Name
mex-3 RNA binding family member D
There are no transcripts associated with this gene.

View Full Product Details