Product Details

SNP ID
rs11812829
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:15097293 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCCGAGCGCTTCCTCCCCGGCCC[C/G]TCGCGTCTGCGAGGCGAGGCCGGGA
Phenotype
MIM: 603801 MIM: 606116
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
ACBD7 PubMed Links
Additional Information
For this assay, SNP(s) [rs117151080] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ACBD7
Gene Name
acyl-CoA binding domain containing 7
There are no transcripts associated with this gene.

Gene
C10orf111
Gene Name
chromosome 10 open reading frame 111
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153244.1 114 UTR 5 NP_694976.1
Gene
NMT2
Gene Name
N-myristoyltransferase 2
There are no transcripts associated with this gene.

Gene
RPP38
Gene Name
ribonuclease P/MRP subunit p38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001097590.2 114 UTR 5 NP_001091059.1
NM_001265601.1 114 UTR 5 NP_001252530.1
NM_006414.4 114 UTR 5 NP_006405.2
NM_183005.4 114 UTR 5 NP_892117.1
XM_006717363.1 114 Intron XP_006717426.1
XM_006717364.3 114 UTR 5 XP_006717427.1
XM_011519293.1 114 Intron XP_011517595.1
XM_017015479.1 114 Intron XP_016870968.1
XM_017015480.1 114 UTR 5 XP_016870969.1
XM_017015481.1 114 Intron XP_016870970.1

View Full Product Details