Product Details

SNP ID
rs901662
Assay Type
Functionally tested
NCBI dbSNP Submissions
68
Location
Chr.1:162381796 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATTTCCAAAGCCTTGGAAGTTACA[A/G]GTTTTGGGTGTGGGATAAGGAAAAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1orf111 PubMed Links

Gene Details

Gene
C1orf111
Gene Name
chromosome 1 open reading frame 111
There are no transcripts associated with this gene.

Gene
C1orf226
Gene Name
chromosome 1 open reading frame 226
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085375.1 67 UTR 5 NP_001078844.1
NM_001135240.1 67 Intron NP_001128712.1

View Full Product Details