Product Details

SNP ID
rs1521177
Assay Type
Functionally tested
NCBI dbSNP Submissions
73
Location
Chr.1:151809654 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTTGAAAGCAGATCCAACCGGAT[G/T]TGCTTTCCAATTGCTCTCCCAGATT
Phenotype
MIM: 609794 MIM: 602943
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LINGO4 PubMed Links

Gene Details

Gene
LINGO4
Gene Name
leucine rich repeat and Ig domain containing 4
There are no transcripts associated with this gene.

Gene
RORC
Gene Name
RAR related orphan receptor C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001523.1 Intron NP_001001523.1
NM_005060.3 Intron NP_005051.2
XM_006711484.3 Intron XP_006711547.2

View Full Product Details