Product Details

SNP ID
rs1016423
Assay Type
Functionally tested
NCBI dbSNP Submissions
65
Location
Chr.1:156644500 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACAGTCCTCCCAAGCACCCCAATG[C/T]CTGGAGCTGGTCTCCGCACCTCTAA
Phenotype
MIM: 600347
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BCAN PubMed Links

Gene Details

Gene
BCAN
Gene Name
brevican
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021948.4 Intron NP_068767.3
NM_198427.1 Intron NP_940819.1
XM_011509866.1 Intron XP_011508168.1
XM_017002047.1 Intron XP_016857536.1

View Full Product Details