Product Details

SNP ID
rs884956
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:122976415 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGATGGTTGAGGTACATCTAGGGG[C/T]TCTCATCCCAGCTAGGCTCAGGTTG
Phenotype
MIM: 605453 MIM: 607668 MIM: 608920
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ABCB9 PubMed Links

Gene Details

Gene
ABCB9
Gene Name
ATP binding cassette subfamily B member 9
There are no transcripts associated with this gene.

Gene
ARL6IP4
Gene Name
ADP ribosylation factor like GTPase 6 interacting protein 4
There are no transcripts associated with this gene.

Gene
OGFOD2
Gene Name
2-oxoglutarate and iron dependent oxygenase domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304833.1 Intron NP_001291762.1
NM_001304834.1 Intron NP_001291763.1
NM_001304835.1 Intron NP_001291764.1
NM_001304836.1 Intron NP_001291765.1
NM_001304837.1 Intron NP_001291766.1
NM_001304838.1 Intron NP_001291767.1
NM_024623.2 Intron NP_078899.1
Gene
PITPNM2
Gene Name
phosphatidylinositol transfer protein membrane associated 2
There are no transcripts associated with this gene.

View Full Product Details