Product Details

SNP ID
rs794558
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:46658924 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTCCCTTTAAGAGGTGGAGCTTAA[C/T]TCTCCTTACCCTTGAATATAGGCTA
Phenotype
MIM: 601501
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
VPS35 PubMed Links

Gene Details

Gene
VPS35
Gene Name
VPS35 retromer complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018206.5 4038 UTR 3 NP_060676.2
XM_005256045.3 4038 Intron XP_005256102.1
XM_011523227.2 4038 Intron XP_011521529.1

View Full Product Details