Product Details

SNP ID
rs8555
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:62163743 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATTATCTTTTTCTTCTTTACCAA[C/T]AGACGATGACAAATGTGGATGGTGA
Phenotype
MIM: 610344
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C2CD4B PubMed Links

Gene Details

Gene
C2CD4B
Gene Name
C2 calcium dependent domain containing 4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007595.2 1712 UTR 3 NP_001007596.2
XM_017022170.1 1712 UTR 3 XP_016877659.1

View Full Product Details