Product Details

SNP ID
rs2977272
Assay Type
Validated
NCBI dbSNP Submissions
53
Location
Chr.1:17210457 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTCAGGGACATTCTGGTCCCCAGCC[C/T]CTGTTTATCTCTTCTGGGAAGACCA
Phenotype
MIM: 607934
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PADI1 PubMed Links

Gene Details

Gene
PADI1
Gene Name
peptidyl arginine deiminase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013358.2 Intron NP_037490.2
XM_011541307.1 Intron XP_011539609.1
XM_017001101.1 Intron XP_016856590.1
XM_017001102.1 Intron XP_016856591.1
XM_017001103.1 Intron XP_016856592.1

View Full Product Details