Product Details

SNP ID
rs1550871
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:18729339 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTCCTTGTCTGCCCATCAGTCCGT[A/G]CCAGTGTTTTCCATCTGCCCCTCAC
Phenotype
MIM: 176879
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IGSF22 PubMed Links

Gene Details

Gene
IGSF22
Gene Name
immunoglobulin superfamily member 22
There are no transcripts associated with this gene.

Gene
PTPN5
Gene Name
protein tyrosine phosphatase, non-receptor type 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039970.1 Intron NP_001035059.1
NM_001278236.1 Intron NP_001265165.1
NM_001278238.1 Intron NP_001265167.1
NM_001278239.1 Intron NP_001265168.1
NM_006906.1 Intron NP_008837.1
NM_032781.3 Intron NP_116170.3
XM_011520411.2 Intron XP_011518713.1
XM_017018434.1 Intron XP_016873923.1
XM_017018435.1 Intron XP_016873924.1
XM_017018436.1 Intron XP_016873925.1
XM_017018437.1 Intron XP_016873926.1
XM_017018438.1 Intron XP_016873927.1
XM_017018439.1 Intron XP_016873928.1
XM_017018440.1 Intron XP_016873929.1
XM_017018441.1 Intron XP_016873930.1

View Full Product Details