Product Details

SNP ID
rs784379
Assay Type
Functionally tested
NCBI dbSNP Submissions
47
Location
Chr.1:211393418 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGATCCATTGTTTTCTTAGATTCAT[A/G]TCTTCCTGTCTCTTAGTGTTCTGCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00467 PubMed Links

Gene Details

Gene
LINC00467
Gene Name
long intergenic non-protein coding RNA 467
There are no transcripts associated with this gene.

View Full Product Details