Product Details

SNP ID
rs905733
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:65660664 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATGTCTAAACTTTCAATTTATTG[G/T]CTGATTTCTGGATATTTCTGTATTT
Phenotype
MIM: 600382 MIM: 603617
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DENND4A PubMed Links

Gene Details

Gene
DENND4A
Gene Name
DENN domain containing 4A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144823.2 7157 UTR 3 NP_001138295.1
NM_001320835.1 7157 UTR 3 NP_001307764.1
NM_005848.4 7157 UTR 3 NP_005839.3
XM_005254120.4 7157 UTR 3 XP_005254177.1
XM_005254121.3 7157 UTR 3 XP_005254178.1
XM_005254122.4 7157 UTR 3 XP_005254179.1
XM_006720371.3 7157 UTR 3 XP_006720434.1
XM_011521156.2 7157 Intron XP_011519458.1
XM_011521157.2 7157 UTR 3 XP_011519459.1
XM_017021862.1 7157 UTR 3 XP_016877351.1
XM_017021863.1 7157 UTR 3 XP_016877352.1
XM_017021864.1 7157 Intron XP_016877353.1
XM_017021865.1 7157 UTR 3 XP_016877354.1
Gene
SLC24A1
Gene Name
solute carrier family 24 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001254740.1 7157 Intron NP_001241669.1
NM_001301031.1 7157 Intron NP_001287960.1
NM_001301032.1 7157 Intron NP_001287961.1
NM_001301033.1 7157 Intron NP_001287962.1
NM_004727.2 7157 Intron NP_004718.1
XM_005254778.3 7157 Intron XP_005254835.1
XM_005254781.3 7157 Intron XP_005254838.1
XM_006720768.3 7157 Intron XP_006720831.1
XM_011522219.1 7157 Intron XP_011520521.1
XM_011522220.2 7157 Intron XP_011520522.1
XM_011522221.2 7157 Intron XP_011520523.1
XM_011522222.2 7157 Intron XP_011520524.1
XM_011522226.2 7157 Intron XP_011520528.1
XM_017022724.1 7157 Intron XP_016878213.1
XM_017022725.1 7157 Intron XP_016878214.1

View Full Product Details