Product Details
- SNP ID
-
rs7759227
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:34760453 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TATTTTTAGTAGCTAGTCAGTTCTT[C/G]CTTATAGGTCCCTAGTCCTTTTTTT
- Phenotype
-
MIM: 603522
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SNRPC
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs16893989] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SNRPC
- Gene Name
- small nuclear ribonucleoprotein polypeptide C
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003093.2 |
|
Intron |
|
|
NP_003084.1 |
View Full Product Details