Product Details

SNP ID
rs34098874
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:33017995 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTCCCATATGCTGTCTCATTAAT[A/G]TATGTGAATTAAAGCAGAGACCAAT
Phenotype
MIM: 612294
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DEPDC7 PubMed Links

Gene Details

Gene
DEPDC7
Gene Name
DEP domain containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077242.1 Intron NP_001070710.1
NM_139160.2 Intron NP_631899.2

View Full Product Details