Product Details
- SNP ID
-
rs11602794
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:3221677 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGTGTCAGCTCAAGCCAGAAATTC[A/G]GTCGTAAAGATGCTGTTTCTCTTCA
- Phenotype
-
MIM: 607232
MIM: 607234
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MRGPRE
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79606774] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MRGPRE
- Gene Name
- MAS related GPR family member E
There are no transcripts associated with this gene.
- Gene
- MRGPRG
- Gene Name
- MAS related GPR family member G
There are no transcripts associated with this gene.
- Gene
- MRGPRG-AS1
- Gene Name
- MRGPRG antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details