Product Details
- SNP ID
-
rs7664769
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:83091864 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTTTCAATTTTCAAATTTTATTCA[A/G]TCATTTATGTCAGCACAAACTCATA
- Phenotype
-
MIM: 607515
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PLAC8
PubMed Links
Gene Details
- Gene
- PLAC8
- Gene Name
- placenta specific 8
View Full Product Details