Product Details
- SNP ID
-
rs6863398
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:75513951 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- TATATTAGGTATCAGTAGAATCCTC[A/T]TGATAGTTATTTAGTAAACGTGTTT
- Phenotype
-
MIM: 604677
MIM: 605650
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
COL4A3BP
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs10072576] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- COL4A3BP
- Gene Name
- collagen type IV alpha 3 binding protein
There are no transcripts associated with this gene.
- Gene
- POLK
- Gene Name
- polymerase (DNA) kappa
View Full Product Details