Product Details

SNP ID
rs35110243
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.15:24581421 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAAATAGACAACGCTTCCAAAAT[C/T]TCTGATATTTAAACCTAGTGGGGAT
Phenotype
MIM: 611215
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
PWRN1 PubMed Links

Gene Details

Gene
PWRN1
Gene Name
Prader-Willi region non-protein coding RNA 1
There are no transcripts associated with this gene.

View Full Product Details