Product Details

SNP ID
rs4074271
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:113651090 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGCTTCGAGGGAAGTGAGGTTCC[C/G]TCGGACACCCTAGTGGGAAGGCTCC
Phenotype
MIM: 137217 MIM: 180381 MIM: 189902
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ATP4B PubMed Links

Gene Details

Gene
ATP4B
Gene Name
ATPase H+/K+ transporting beta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000705.3 Intron NP_000696.1
Gene
GRK1
Gene Name
G protein-coupled receptor kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002929.2 Intron NP_002920.1
XM_017020684.1 Intron XP_016876173.1
Gene
TFDP1
Gene Name
transcription factor Dp-1
There are no transcripts associated with this gene.

View Full Product Details