Product Details

SNP ID
rs12550827
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:124542846 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTTTTTGTTTAAATCCTCATTCC[A/C]TTGTTGGATGTAAGAGGAGAAAAAT
Phenotype
MIM: 608486 MIM: 601445
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MTSS1 PubMed Links

Gene Details

Gene
MTSS1
Gene Name
metastasis suppressor 1
There are no transcripts associated with this gene.

Gene
NDUFB9
Gene Name
NADH:ubiquinone oxidoreductase subunit B9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278645.1 Intron NP_001265574.1
NM_001278646.1 Intron NP_001265575.1
NM_001311168.1 Intron NP_001298097.1
NM_005005.2 Intron NP_004996.1
Gene
TATDN1
Gene Name
TatD DNase domain containing 1
There are no transcripts associated with this gene.

View Full Product Details