Product Details
- SNP ID
-
rs2027330
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
39
- Location
-
Chr.1:10215534 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CAAAGCATATCATTCAACCATACTA[C/T]CCTCTACCATTGCCCGTTTTTTTCC
- Phenotype
-
MIM: 605995
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KIF1B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115686695] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KIF1B
- Gene Name
- kinesin family member 1B
View Full Product Details