Product Details

SNP ID
rs11771715
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:139342750 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGTTGTAATTACAGGCGTGAACTA[C/T]CAAACCTGGCCCCATTCTAAGATTT
Phenotype
MIM: 613056
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C7orf55-LUC7L2 PubMed Links

Gene Details

Gene
C7orf55-LUC7L2
Gene Name
C7orf55-LUC7L2 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244584.2 Intron NP_001231513.1
Gene
FMC1
Gene Name
formation of mitochondrial complex V assembly factor 1 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_197964.4 Intron NP_932068.2
Gene
LUC7L2
Gene Name
LUC7 like 2, pre-mRNA splicing factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244585.1 Intron NP_001231514.1
NM_001270643.1 Intron NP_001257572.1
NM_016019.4 Intron NP_057103.2

View Full Product Details