Product Details

SNP ID
rs10426212
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:14603764 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACAGGAGTGAGCCACCGTGCCCTA[A/G]CCCATAGCTCACTTCTTAATTCATT
Phenotype
MIM: 606101 MIM: 616838
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADGRE3 PubMed Links

Gene Details

Gene
ADGRE3
Gene Name
adhesion G protein-coupled receptor E3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289158.1 Intron NP_001276087.1
NM_001289159.1 Intron NP_001276088.1
NM_032571.4 Intron NP_115960.2
XM_011528374.2 Intron XP_011526676.1
XM_017027383.1 Intron XP_016882872.1
Gene
CLEC17A
Gene Name
C-type lectin domain family 17 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204118.1 Intron NP_001191047.1
NM_207390.3 Intron NP_997273.3
XM_017026785.1 Intron XP_016882274.1
XM_017026786.1 Intron XP_016882275.1
XM_017026787.1 Intron XP_016882276.1
XM_017026788.1 Intron XP_016882277.1
XM_017026789.1 Intron XP_016882278.1
XM_017026790.1 Intron XP_016882279.1
XM_017026791.1 Intron XP_016882280.1
XM_017026792.1 Intron XP_016882281.1
XM_017026793.1 Intron XP_016882282.1
XM_017026794.1 Intron XP_016882283.1

View Full Product Details