Product Details

SNP ID
rs2240927
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:9251667 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTCAACTCCTCCACCTTAGGTGTTC[C/T]GACACCTTCATCAATGAAATGGTCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR7E18P PubMed Links

Gene Details

Gene
OR7E18P
Gene Name
olfactory receptor family 7 subfamily E member 18 pseudogene
There are no transcripts associated with this gene.

Gene
OR7E24
Gene Name
olfactory receptor family 7 subfamily E member 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079935.1 Intron NP_001073404.1

View Full Product Details