Product Details

SNP ID
rs3923739
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:121817847 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTAAGCCTCCTGGAAGCCACTTCCC[C/T]GCCTGGGAAAACAGAGTGCCAGTGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC107987125 PubMed Links

Gene Details

Gene
LOC107987125
Gene Name
uncharacterized LOC107987125
There are no transcripts associated with this gene.

Gene
TTLL11
Gene Name
tubulin tyrosine ligase like 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001139442.1 6632 Intron NP_001132914.1
NM_194252.2 6632 Intron NP_919228.2
XM_005251728.2 6632 UTR 3 XP_005251785.1
XM_005251729.4 6632 UTR 3 XP_005251786.1
XM_006716966.3 6632 Intron XP_006717029.1
XM_017014309.1 6632 Intron XP_016869798.1

View Full Product Details