Product Details

SNP ID
rs3212
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49108308 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAAATCATACCCCTCAGCTTCCCA[C/T]TGACAGAGCCAGTGTCCTCTGGGTT
Phenotype
MIM: 603727 MIM: 614471
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR6890 PubMed Links

Gene Details

Gene
MIR6890
Gene Name
microRNA 6890
There are no transcripts associated with this gene.

Gene
QARS
Gene Name
glutaminyl-tRNA synthetase
There are no transcripts associated with this gene.

Gene
USP19
Gene Name
ubiquitin specific peptidase 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199160.1 4522 Intron NP_001186089.1
NM_001199161.1 4522 UTR 3 NP_001186090.1
NM_001199162.1 4522 UTR 3 NP_001186091.1
NM_006677.2 4522 Intron NP_006668.1
XM_005264823.2 4522 Intron XP_005264880.1
XM_005264824.2 4522 Intron XP_005264881.1
XM_005264825.2 4522 Intron XP_005264882.1
XM_005264826.2 4522 Intron XP_005264883.1
XM_005264827.2 4522 Intron XP_005264884.1
XM_005264829.2 4522 UTR 3 XP_005264886.1
XM_005264830.2 4522 Intron XP_005264887.1
XM_005264831.2 4522 UTR 3 XP_005264888.1
XM_006712946.2 4522 Intron XP_006713009.1
XM_006712947.2 4522 Intron XP_006713010.1
XM_006712948.2 4522 UTR 3 XP_006713011.1
XM_006712949.2 4522 Intron XP_006713012.1
XM_006712950.2 4522 UTR 3 XP_006713013.1
XM_006712951.2 4522 UTR 3 XP_006713014.1
XM_006712952.2 4522 UTR 3 XP_006713015.1
XM_006712953.1 4522 Intron XP_006713016.1
XM_017005615.1 4522 Intron XP_016861104.1
XM_017005616.1 4522 Intron XP_016861105.1
XM_017005617.1 4522 Intron XP_016861106.1
XM_017005618.1 4522 UTR 3 XP_016861107.1
XM_017005619.1 4522 UTR 3 XP_016861108.1
XM_017005620.1 4522 UTR 3 XP_016861109.1
XM_017005621.1 4522 Intron XP_016861110.1
XM_017005622.1 4522 UTR 3 XP_016861111.1
XM_017005623.1 4522 Intron XP_016861112.1
XM_017005624.1 4522 UTR 3 XP_016861113.1
XM_017005625.1 4522 Intron XP_016861114.1
XM_017005626.1 4522 Intron XP_016861115.1
XM_017005627.1 4522 Intron XP_016861116.1
XM_017005628.1 4522 Intron XP_016861117.1
XM_017005629.1 4522 Intron XP_016861118.1
XM_017005630.1 4522 UTR 3 XP_016861119.1
XM_017005631.1 4522 UTR 3 XP_016861120.1
XM_017005632.1 4522 UTR 3 XP_016861121.1
XM_017005633.1 4522 UTR 3 XP_016861122.1
XM_017005634.1 4522 UTR 3 XP_016861123.1
XM_017005635.1 4522 Intron XP_016861124.1

View Full Product Details