Product Details

SNP ID
rs10485186
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:116497897 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATTGATGTTTATTGAATCTTACCT[A/G]ACTCAGTCCCCTCTCTCACTAAGAA
Phenotype
MIM: 614137
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRAPPC3L PubMed Links

Gene Details

Gene
TRAPPC3L
Gene Name
trafficking protein particle complex 3 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001139444.2 Intron NP_001132916.1

View Full Product Details