Product Details

SNP ID
rs1522257
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:174442372 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATTATTTTAACAATACTTTCACC[G/T]TTAATTTTTTAGAGATTTTTTTTCT
Phenotype
MIM: 608806
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NAALADL2 PubMed Links

Gene Details

Gene
NAALADL2
Gene Name
N-acetylated alpha-linked acidic dipeptidase-like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207015.2 Intron NP_996898.2
XM_006713560.2 Intron XP_006713623.1
XM_011512612.2 Intron XP_011510914.1
XM_011512613.1 Intron XP_011510915.1
XM_011512615.2 Intron XP_011510917.1
XM_011512616.2 Intron XP_011510918.1
XM_011512617.2 Intron XP_011510919.1
XM_017006070.1 Intron XP_016861559.1
XM_017006071.1 Intron XP_016861560.1
XM_017006072.1 Intron XP_016861561.1
XM_017006073.1 Intron XP_016861562.1
XM_017006074.1 Intron XP_016861563.1
XM_017006075.1 Intron XP_016861564.1
XM_017006076.1 Intron XP_016861565.1
XM_017006077.1 Intron XP_016861566.1
XM_017006078.1 Intron XP_016861567.1
XM_017006079.1 Intron XP_016861568.1
XM_017006080.1 Intron XP_016861569.1
XM_017006081.1 Intron XP_016861570.1
XM_017006082.1 Intron XP_016861571.1
XM_017006083.1 Intron XP_016861572.1

View Full Product Details