Product Details

SNP ID
rs36080069
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:13675396 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACATTCAGGGATAAGGTGAGGGT[A/G]ATCACATGGATAAAGCGTGCGGTTA
Phenotype
MIM: 616107
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAR1 PubMed Links

Gene Details

Gene
FAR1
Gene Name
fatty acyl-CoA reductase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032228.5 Intron NP_115604.1
XM_011520400.1 Intron XP_011518702.1

View Full Product Details