Product Details
- SNP ID
-
rs13005305
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:174076872 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTTTTTATAAAGACATGCAATGTAT[G/T]GAAGGCCATAAGCTACTTATTTCTA
- Phenotype
-
MIM: 611175
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
OLA1
PubMed Links
Gene Details
- Gene
- OLA1
- Gene Name
- Obg-like ATPase 1
There are no transcripts associated with this gene.
View Full Product Details