Product Details

SNP ID
rs2687078
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:99707089 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
ACCTTGGAGGGGCTTGGGAATTGAA[C/T]TAGGACTTGAAAGATAAGAACAGAA
Phenotype
MIM: 605340
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYP3A7 PubMed Links

Gene Details

Gene
CYP3A7
Gene Name
cytochrome P450 family 3 subfamily A member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000765.4 Intron NP_000756.3
Gene
CYP3A7-CYP3A51P
Gene Name
CYP3A7-CYP3A51P readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256497.2 Intron NP_001243426.2
Gene
ZSCAN25
Gene Name
zinc finger and SCAN domain containing 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145115.2 Intron NP_660090.2
XM_005250194.2 Intron XP_005250251.1
XM_011515905.2 Intron XP_011514207.1
XM_011515906.2 Intron XP_011514208.1
XM_011515907.2 Intron XP_011514209.1
XM_011515908.2 Intron XP_011514210.1
XM_011515909.2 Intron XP_011514211.1
XM_011515910.2 Intron XP_011514212.1
XM_017011824.1 Intron XP_016867313.1
XM_017011825.1 Intron XP_016867314.1
XM_017011826.1 Intron XP_016867315.1
XM_017011827.1 Intron XP_016867316.1

View Full Product Details