Product Details

SNP ID
rs7634165
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:189601310 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATGAGCACATGTGCTCATTTACT[C/G]TCCTTCAGAAATGTCCACATCTAGC
Phenotype
MIM: 603273
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TP63 PubMed Links

Gene Details

Gene
TP63
Gene Name
tumor protein p63
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114978.1 Intron NP_001108450.1
NM_001114979.1 Intron NP_001108451.1
NM_001114980.1 Intron NP_001108452.1
NM_001114981.1 Intron NP_001108453.1
NM_001114982.1 Intron NP_001108454.1
NM_003722.4 Intron NP_003713.3
XM_005247843.3 Intron XP_005247900.1
XM_005247844.4 Intron XP_005247901.1
XM_011513251.2 Intron XP_011511553.1
XM_011513252.2 Intron XP_011511554.1
XM_017007387.1 Intron XP_016862876.1

View Full Product Details