Product Details

SNP ID
rs11932822
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:155175287 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGTATAATCTGCTGAAGTATTTG[C/T]AAATACACAGGGGAGATACAGTCAG
Phenotype
MIM: 162642
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NPY2R PubMed Links

Gene Details

Gene
NPY2R
Gene Name
neuropeptide Y receptor Y2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000910.3 Intron NP_000901.1
XM_005263033.4 Intron XP_005263090.1
XM_005263034.4 Intron XP_005263091.1

View Full Product Details