Product Details

SNP ID
rs10920570
Assay Type
Functionally tested
NCBI dbSNP Submissions
48
Location
Chr.1:203129690 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCAAAGGCACGCTTTGGCTCTCTC[A/G]TTCACCAGGTATCTCTGTGCTAGCC
Phenotype
MIM: 102775
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADORA1 PubMed Links

Gene Details

Gene
ADORA1
Gene Name
adenosine A1 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000674.2 Intron NP_000665.1
NM_001048230.1 Intron NP_001041695.1
XM_005244901.1 Intron XP_005244958.1
XM_005244902.4 Intron XP_005244959.1
XM_017000342.1 Intron XP_016855831.1

View Full Product Details