Product Details

SNP ID
rs11902796
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:26350654 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATAACTCAGTTTGGTGGCATGTGA[A/G]CATGTAAATGAACAGTTATAAAAAG
Phenotype
MIM: 607915
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADGRF3 PubMed Links

Gene Details

Gene
ADGRF3
Gene Name
adhesion G protein-coupled receptor F3
There are no transcripts associated with this gene.

Gene
EPT1
Gene Name
ethanolaminephosphotransferase 1
There are no transcripts associated with this gene.

View Full Product Details