Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001007257.2 | Intron | NP_001007258.1 | |||
NM_001135610.1 | Intron | NP_001129082.1 | |||
NM_012231.4 | Intron | NP_036363.2 | |||
NM_015866.4 | Intron | NP_056950.2 | |||
XM_017002253.1 | Intron | XP_016857742.1 | |||
XM_017002254.1 | Intron | XP_016857743.1 | |||
XM_017002255.1 | Intron | XP_016857744.1 | |||
XM_017002256.1 | Intron | XP_016857745.1 | |||
XM_017002257.1 | Intron | XP_016857746.1 | |||
XM_017002258.1 | Intron | XP_016857747.1 | |||
XM_017002259.1 | Intron | XP_016857748.1 | |||
XM_017002260.1 | Intron | XP_016857749.1 | |||
XM_017002261.1 | Intron | XP_016857750.1 | |||
XM_017002262.1 | Intron | XP_016857751.1 | |||
XM_017002263.1 | Intron | XP_016857752.1 | |||
XM_017002264.1 | Intron | XP_016857753.1 |