Product Details

SNP ID
rs35106872
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:21494823 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACTAGGTGGGCCGCTCGGCCCCA[A/G]GCCTGGCAGGGCGGAGCCTAGGCCG
Phenotype
MIM: 615202
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CASC10 PubMed Links

Gene Details

Gene
CASC10
Gene Name
cancer susceptibility candidate 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010911.2 1658 UTR 3 NP_001010911.1
Gene
MIR1915
Gene Name
microRNA 1915
There are no transcripts associated with this gene.

View Full Product Details