Product Details

SNP ID
rs11983581
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:50465190 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCATAAAAGGCAATATGTTGTATG[A/T]TTTCATTTATACAAATGTCCAGAAT
Phenotype
MIM: 107930
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DDC PubMed Links

Gene Details

Gene
DDC
Gene Name
dopa decarboxylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000790.3 Intron NP_000781.1
NM_001082971.1 Intron NP_001076440.1
NM_001242886.1 Intron NP_001229815.1
NM_001242887.1 Intron NP_001229816.1
NM_001242888.1 Intron NP_001229817.1
NM_001242889.1 Intron NP_001229818.1
NM_001242890.1 Intron NP_001229819.1
XM_005271745.4 Intron XP_005271802.1
XM_011515161.2 Intron XP_011513463.2

View Full Product Details