Product Details
- SNP ID
-
rs2722191
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:103952179 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGAAACCCCATACTTATTAGCAGTC[A/C]CTTTCATTTCCATACAACCCTCCCC
- Phenotype
-
MIM: 191175
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
C12orf73
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79579452] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C12orf73
- Gene Name
- chromosome 12 open reading frame 73
- Gene
- HSP90B1
- Gene Name
- heat shock protein 90 beta family member 1
There are no transcripts associated with this gene.
View Full Product Details