Product Details

SNP ID
rs470086
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.22:43826559 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TCACAGGCAGCCCTGGCCCTTCCCA[C/T]GCCATTATCCACTCACAGCTCAACA
Phenotype
MIM: 608359
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105373057 PubMed Links

Gene Details

Gene
LOC105373057
Gene Name
DNA-directed RNA polymerase II subunit RPB1-like
There are no transcripts associated with this gene.

Gene
SULT4A1
Gene Name
sulfotransferase family 4A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014351.3 Intron NP_055166.1
XM_011530120.2 Intron XP_011528422.1
XM_011530121.1 Intron XP_011528423.1

View Full Product Details