Product Details

SNP ID
rs3801916
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:95491707 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATCAGCATCAGCAGAAGATTGTGAA[C/T]AGGCACAGCCTCATCTGTAGTTTTT
Phenotype
MIM: 605761
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ASB4 PubMed Links

Gene Details

Gene
ASB4
Gene Name
ankyrin repeat and SOCS box containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016116.2 Intron NP_057200.1
NM_145872.2 Intron NP_665879.1
XM_017012303.1 Intron XP_016867792.1

View Full Product Details