Product Details

SNP ID
rs4947963
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:55020722 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCTTGGAAGGTTTAATTGCACAAT[C/T]CCAACCTTGAGCTGCGCGGGTTCCA
Phenotype
MIM: 131550
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EGFR PubMed Links

Gene Details

Gene
EGFR
Gene Name
epidermal growth factor receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005228.3 Intron NP_005219.2
NM_201282.1 Intron NP_958439.1
NM_201283.1 Intron NP_958440.1
NM_201284.1 Intron NP_958441.1

View Full Product Details