Product Details
- SNP ID
-
rs5760747
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:24938902 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TATACTGCACAACTCTCAGAGACAC[A/G]TTGGGGGCCATCATAGATTTGCAAA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM211
PubMed Links
Gene Details
- Gene
- TMEM211
- Gene Name
- transmembrane protein 211
View Full Product Details