Product Details

SNP ID
rs7300366
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6543061 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGCCACCTTGAAAACTGCAGGCTC[A/G]CTGTGTGAACAGACTTGGGGGTGGG
Phenotype
MIM: 138400 MIM: 610495
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GAPDH PubMed Links

Gene Details

Gene
GAPDH
Gene Name
glyceraldehyde-3-phosphate dehydrogenase
There are no transcripts associated with this gene.

Gene
IFFO1
Gene Name
intermediate filament family orphan 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039670.2 Intron NP_001034759.1
NM_001193457.1 Intron NP_001180386.1
NM_080730.4 Intron NP_542768.2
XM_006718975.3 Intron XP_006719038.1
XM_006718976.3 Intron XP_006719039.1
XM_006718977.3 Intron XP_006719040.1
XM_006718981.3 Intron XP_006719044.1
XM_011520947.2 Intron XP_011519249.1
XM_011520948.2 Intron XP_011519250.1
XM_011520949.2 Intron XP_011519251.1
XM_011520952.2 Intron XP_011519254.1
XM_017019169.1 Intron XP_016874658.1
XM_017019170.1 Intron XP_016874659.1
XM_017019171.1 Intron XP_016874660.1
XM_017019172.1 Intron XP_016874661.1
XM_017019173.1 Intron XP_016874662.1

View Full Product Details