Product Details

SNP ID
rs2058215
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:76732000 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CAACTGGAAAATCTTTACTAGAAAG[C/T]CATGTTGCAGACCTGTTAGGTTTTA
Phenotype
MIM: 604914 MIM: 615262 MIM: 600813
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
JMJD6 PubMed Links

Gene Details

Gene
JMJD6
Gene Name
arginine demethylase and lysine hydroxylase
There are no transcripts associated with this gene.

Gene
METTL23
Gene Name
methyltransferase like 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080510.4 Intron NP_001073979.3
NM_001206983.2 Intron NP_001193912.1
NM_001206984.2 Intron NP_001193913.1
NM_001206985.2 Intron NP_001193914.1
NM_001206986.2 Intron NP_001193915.1
NM_001206987.2 Intron NP_001193916.1
NM_001302703.1 Intron NP_001289632.1
NM_001302704.1 Intron NP_001289633.1
NM_001302705.1 Intron NP_001289634.1
XM_006721674.3 Intron XP_006721737.1
XM_006721675.1 Intron XP_006721738.1
XM_006721676.3 Intron XP_006721739.1
XM_006721678.3 Intron XP_006721741.1
XM_006721679.3 Intron XP_006721742.1
XM_006721680.2 Intron XP_006721743.1
XM_011524282.1 Intron XP_011522584.1
XM_017024145.1 Intron XP_016879634.1
XM_017024146.1 Intron XP_016879635.1
Gene
MFSD11
Gene Name
major facilitator superfamily domain containing 11
There are no transcripts associated with this gene.

Gene
MIR636
Gene Name
microRNA 636
There are no transcripts associated with this gene.

Gene
SRSF2
Gene Name
serine and arginine rich splicing factor 2
There are no transcripts associated with this gene.

View Full Product Details