Product Details
- SNP ID
-
rs6510624
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:1783795 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTCTGGGGCCCATCTCAGCTGCAAC[C/T]TGCTTTTCCATGCATGCTTTGGTTA
- Phenotype
-
MIM: 605866
MIM: 611294
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ATP8B3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs141226344] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ATP8B3
- Gene Name
- ATPase phospholipid transporting 8B3
- Gene
- ONECUT3
- Gene Name
- one cut homeobox 3
There are no transcripts associated with this gene.
View Full Product Details