Product Details
- SNP ID
-
rs11189412
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:97851956 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCCTGTGTACAAAGCACTATCGCAT[A/G]AGTGATCTCTTTCAAGCCTCAGAGC
- Phenotype
-
MIM: 606276
MIM: 614189
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CRTAC1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs139719041] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CRTAC1
- Gene Name
- cartilage acidic protein 1
There are no transcripts associated with this gene.
- Gene
- GOLGA7B
- Gene Name
- golgin A7 family member B
- Gene
- LINC00866
- Gene Name
- long intergenic non-protein coding RNA 866
There are no transcripts associated with this gene.
View Full Product Details