Product Details

SNP ID
rs12546625
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:120142986 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGTCATGTTACTTTGAAAGTTTAC[A/G]TAACTATGCTTTGCTGCATTCATCA
Phenotype
MIM: 120324
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COL14A1 PubMed Links

Gene Details

Gene
COL14A1
Gene Name
collagen type XIV alpha 1 chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021110.3 Intron NP_066933.1
XM_005251059.3 Intron XP_005251116.1
XM_006716651.3 Intron XP_006716714.1
XM_017013809.1 Intron XP_016869298.1

View Full Product Details