Product Details

SNP ID
rs2416937
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:99918355 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATGCTGCCCAGGCCTATCTCATCT[A/C]AAAGCCCTGGCCTCAAGTGATCCTC
Phenotype
MIM: 604204
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
STX17 PubMed Links

Gene Details

Gene
STX17
Gene Name
syntaxin 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017919.2 Intron NP_060389.2
XM_011518820.2 Intron XP_011517122.1
XM_011518821.2 Intron XP_011517123.1
XM_011518823.2 Intron XP_011517125.1
XM_017014875.1 Intron XP_016870364.1

View Full Product Details