Product Details

SNP ID
rs34765154
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:28162672 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCTGGTTCCAAGGATTCTGTTAAT[A/G]AGAAAACTCCACAGGCCAGCATTCT
Phenotype
MIM: 602240
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZKSCAN8 PubMed Links

Gene Details

Gene
ZKSCAN8
Gene Name
zinc finger with KRAB and SCAN domains 8
There are no transcripts associated with this gene.

Gene
ZNF192P1
Gene Name
zinc finger protein 192 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details